accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 102-79-4

IUPAC Name: 2-[butyl(2-hydroxyethyl)amino]ethan-1-ol
Synonyms: butyl diethanolamine
Molecular Weight (g/mol): 161.25
Molecular Formula: C8H19NO2
InChi Key: GVNHOISKXMSMPX-UHFFFAOYSA-N
SMILES: CCCCN(CCO)CCO
Boiling Point 
  • (3)
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (5)
  • (4)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)