accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 102-81-8

IUPAC Name: 2-(dibutylamino)ethan-1-ol
Synonyms: 2-dibutylaminoethanol dibutyl ethanolamine ethanolamine,N,N-dibutyl
Molecular Weight (g/mol): 173.3
Molecular Formula: C10H23NO
InChi Key: IWSZDQRGNFLMJS-UHFFFAOYSA-N
SMILES: CCCCN(CCO)CCCC
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)