accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 102-83-0

IUPAC Name: (3-aminopropyl)dibutylamine
Synonyms: (3-aminopropyl)dibutylamine
Molecular Weight (g/mol): 186.34
Molecular Formula: C11H26N2
InChi Key: KYCGURZGBKFEQB-UHFFFAOYSA-N
SMILES: CCCCN(CCCC)CCCN
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)