accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH2OPOCH2OCH2

CAS RN 102-84-1

IUPAC Name: tris(prop-2-en-1-yl) phosphite
Synonyms: tris(prop-2-en-1-yl) phosphite
Molecular Weight (g/mol): 202.19
Molecular Formula: C9H15O3P
InChi Key: KJWHEZXBZQXVSA-UHFFFAOYSA-N
SMILES: C=CCOP(OCC=C)OCC=C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)