accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NO

CAS RN 102-93-2

IUPAC Name: 3-phenylpropanamide
Synonyms: propionamide, G-phenyl
Molecular Weight (g/mol): 149.19
Molecular Formula: C9H11NO
InChi Key: VYIBCOSBNVFEIW-UHFFFAOYSA-N
SMILES: NC(=O)CCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results