accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiH3CNHSiCH3CH3NHSiCH3H3CHNSiH3CH3CHN

CAS RN 1020-84-4

IUPAC Name: 2,2,4,4,6,6,8,8-octamethyl-1,3,5,7,2,4,6,8-tetrazatetrasilocane
Synonyms: 2,2,4,4,6,6,8,8-octamethyl-1,3,5,7,2,4,6,8-tetrazatetrasilocane
Molecular Weight (g/mol): 292.68
Molecular Formula: C8H28N4Si4
InChi Key: FIADVASZMLCQIF-UHFFFAOYSA-N
SMILES: C[Si]1(C)N[Si](C)(C)N[Si](C)(C)N[Si](C)(C)N1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)