accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1020056-98-7

NOOHBrBr

CAS RN 1020056-98-7

IUPAC Name: 2,3-dibromopyridine-4-carboxylic acid
Synonyms: 2,3-dibromopyridine-4-carboxylic acid
Molecular Weight (g/mol): 280.9
Molecular Formula: C6H3Br2NO2
InChi Key: ICISUZSRQHUJKX-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(Br)C(Br)=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results