CAS RN 10202-45-6
IUPAC Name:
2-{[1,1'-biphenyl]-4-yl}-4,6-dichloro-1,3,5-triazine
Synonyms:
2-{[1,1'-biphenyl]-4-yl}-4,6-dichloro-1,3,5-triazine
Molecular Weight (g/mol):
302.16
Molecular Formula:
C15H9Cl2N3
InChi Key:
JYPGHMDTTDKUEL-UHFFFAOYSA-N
SMILES:
ClC1=NC(=NC(Cl)=N1)C1=CC=C(C=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
2-(4-Biphenylyl)-4,6-dichloro-1,3,5-triazine 97.0+%, TCI America™
CAS: 10202-45-6 Molecular Formula: C15H9Cl2N3 Molecular Weight (g/mol): 302.158 InChI Key: JYPGHMDTTDKUEL-UHFFFAOYSA-N PubChem CID: 22352243 IUPAC Name: 2,4-dichloro-6-(4-phenylphenyl)-1,3,5-triazine SMILES: C1=CC=C(C=C1)C2=CC=C(C=C2)C3=NC(=NC(=N3)Cl)Cl