accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3OHHNSOOOHONa

CAS RN 102029-60-7

IUPAC Name: sodium 2-hydroxy-3-[(1-hydroxy-2-methylpropan-2-yl)amino]propane-1-sulfonate
Synonyms: sodium 2-hydroxy-3-[(1-hydroxy-2-methylpropan-2-yl)amino]propane-1-sulfonate
Molecular Weight (g/mol): 249.26
Molecular Formula: C7H16NNaO5S
InChi Key: UVGYCHPTWHWEPZ-UHFFFAOYNA-M
SMILES: [Na+].CC(C)(CO)NCC(O)CS([O-])(=O)=O
Quantity 
  • (2)
  • (1)
pH 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)