CAS RN 102065-89-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
3-Aminoazetidine dihydrochloride, 95%
3-Aminoazetidine dihydrochloride, 95%
CAS: 102065-89-4 Molecular Formula: C3H8N2 Molecular Weight (g/mol): 72.111 MDL Number: MFCD09910173 InChI Key: FDPKMJDUXJFKOI-UHFFFAOYSA-N Synonym: 3-aminoazetidine,3-azetidinamine,acmc-209y1n,3-aminoazetidine dihy drochloric PubChem CID: 1516506 IUPAC Name: azetidin-3-amine SMILES: C1C(CN1)N