CAS RN 1020719-61-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-(Methyl-d3-nitrosamino)-1-(3-pyridyl)-1-butanol, ≥97% (HPLC), MilliporeSigma™ Supelco™
Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706031, T706021