accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNSOOCl

CAS RN 1020721-61-2

IUPAC Name: 1-methyl-1H-pyrazole-5-sulfonyl chloride
Synonyms: 2-methylpyrazole-3-sulfonyl chloride
Molecular Weight (g/mol): 180.61
Molecular Formula: C4H5ClN2O2S
InChi Key: OWLKLUGPVSESBX-UHFFFAOYSA-N
SMILES: CN1N=CC=C1S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results