accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 102127-34-4

H3COHOBr

CAS RN 102127-34-4

IUPAC Name: 4-bromo-3-methoxyphenol
Synonyms: 4-bromo-3-methoxyphenol
Molecular Weight (g/mol): 203.04
Molecular Formula: C7H7BrO2
InChi Key: UYDZUCNMZXCLJI-UHFFFAOYSA-N
SMILES: COC1=C(Br)C=CC(O)=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results