accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOHNNOHOHNOOCH3CH3H3CNH2H3CH3COH3C(E/Z)

CAS RN 102185-38-6

IUPAC Name: 5-{[amino(4-methoxy-2,3,6-trimethylbenzenesulfonamido)methylidene]amino}-2-{[(tert-butoxy)carbonyl]amino}pentanoic acid
Synonyms: 5-{[amino(4-methoxy-2,3,6-trimethylbenzenesulfonamido)methylidene]amino}-2-[(tert-butoxycarbonyl)amino]pentanoic acid
Molecular Weight (g/mol): 486.58
Molecular Formula: C21H34N4O7S
InChi Key: CXZHJRGYWGPJSD-UHFFFAOYNA-N
SMILES: COC1=C(C)C(C)=C(C(C)=C1)S(=O)(=O)NC(N)=NCCCC(NC(=O)OC(C)(C)C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results