accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 102187-19-9

IUPAC Name: 4-{[4-(diethylamino)-2-methylphenyl]imino}-1-oxo-N-phenyl-1,4-dihydronaphthalene-2-carboxamide
Synonyms: 4-{[4-(diethylamino)-2-methylphenyl]imino}-1-oxo-N-phenylnaphthalene-2-carboxamide
Molecular Weight (g/mol): 437.54
Molecular Formula: C28H27N3O2
InChi Key: ZYKBEIDPRRYKKQ-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC(C)=C(C=C1)N=C1C=C(C(=O)NC2=CC=CC=C2)C(=O)C2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results