accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1022-79-3

IUPAC Name: 4-amino-5-bromo-1-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one
Synonyms: 4-amino-5-bromo-1-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
Molecular Weight (g/mol): 306.12
Molecular Formula: C9H12BrN3O4
InChi Key: KISUPFXQEHWGAR-UHFFFAOYNA-N
SMILES: NC1=NC(=O)N(C=C1Br)C1CC(O)C(CO)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)