accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHNHCl

CAS RN 1022094-03-6

IUPAC Name: 3-methylmorpholine hydrochloride
Synonyms: 3-methylmorpholine hydrochloride
Molecular Weight (g/mol): 137.61
Molecular Formula: C5H12ClNO
InChi Key: MSOCQCWIEBVSLF-UHFFFAOYNA-N
SMILES: Cl.CC1COCCN1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)