accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10222-01-2

IUPAC Name: 2,2-dibromo-2-cyanoacetamide
Synonyms: dbnpa
Molecular Weight (g/mol): 241.87
Molecular Formula: C3H2Br2N2O
InChi Key: UUIVKBHZENILKB-UHFFFAOYSA-N
SMILES: NC(=O)C(Br)(Br)C#N
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results