accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 102269-42-1

IUPAC Name: 3-(furan-2-yl)aniline
Synonyms: 3-(furan-2-yl)aniline
Molecular Weight (g/mol): 159.19
Molecular Formula: C10H9NO
InChi Key: SLNKACMTMZYMNA-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=C1)C1=CC=CO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results