accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOFFFClHOOH

CAS RN 1022922-16-2

IUPAC Name: [2-chloro-5-(trifluoromethoxy)phenyl]boronic acid
Synonyms: 2-chloro-5-(trifluoromethoxy)phenylboronic acid
Molecular Weight (g/mol): 240.37
Molecular Formula: C7H5BClF3O3
InChi Key: BAEHYECKXGVYKR-UHFFFAOYSA-N
SMILES: OB(O)C1=C(Cl)C=CC(OC(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results