accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 102297-41-6

IUPAC Name: tert-butyl N-[(pyridin-3-yl)methyl]carbamate
Synonyms: tert-butyl N-(pyridin-3-ylmethyl)carbamate
Molecular Weight (g/mol): 208.26
Molecular Formula: C11H16N2O2
InChi Key: PHQXGCNECRISKB-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC1=CN=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)