accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SHSHH3C

CAS RN 10230-61-2

IUPAC Name: [4,5-dimethyl-2-(sulfanylmethyl)phenyl]methanethiol
Synonyms: [4,5-dimethyl-2-(sulfanylmethyl)phenyl]methanethiol
Molecular Weight (g/mol): 198.34
Molecular Formula: C10H14S2
InChi Key: NNGQXIUDFKIVKI-UHFFFAOYSA-N
SMILES: CC1=CC(CS)=C(CS)C=C1C
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results