accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHNSNH2

CAS RN 102353-42-4

IUPAC Name: (2-methoxyethyl)thiourea
Synonyms: 2-methoxyethylthiourea
Molecular Weight (g/mol): 134.2
Molecular Formula: C4H10N2OS
InChi Key: XLJXJKHWLMYXBE-UHFFFAOYSA-N
SMILES: COCCNC(N)=S
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)