accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10236-47-2

IUPAC Name: (2S)-7-{[(2S,3R,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-{[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}oxan-2-yl]oxy}-5-hydroxy-2-(4-hydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-4-one
Synonyms: naringin naringin hydrate aurantiin naringoside cyclorel 4'5-dioh-flavone-7-rhgluc naringenin-7-hesperidoside alpharma brand of naringin sodium naringin sodium
Molecular Weight (g/mol): 580.54
Molecular Formula: C27H32O14
InChi Key: DFPMSGMNTNDNHN-ZPHOTFPESA-N
SMILES: C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2OC2=CC(O)=C3C(=O)C[C@H](OC3=C2)C2=CC=C(O)C=C2)[C@H](O)[C@H](O)[C@H]1O
Percent Purity 
  • (1)
  • (1)
  • (1)
  • (2)
Quantity 
  • (2)
  • (3)
Physical Form 
  • (1)
  • (2)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)