accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOOHNFFNOO

CAS RN 1023805-47-1

IUPAC Name: N-(2,6-difluorophenyl)-3-nitrobenzene-1-sulfonamide
Synonyms: N-(2,6-difluorophenyl)-3-nitrobenzenesulfonamide
Molecular Weight (g/mol): 314.26
Molecular Formula: C12H8F2N2O4S
InChi Key: MTBZLMOKCNKJCR-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC=C1)S(=O)(=O)NC1=C(F)C=CC=C1F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results