accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNHOO

CAS RN 10241-97-1

IUPAC Name: 5-methyl-1H-indole-2-carboxylic acid
Synonyms: 5-methyl-1H-indole-2-carboxylic acid
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: DAITVOCMWPNFTL-UHFFFAOYSA-N
SMILES: CC1=CC=C2NC(=CC2=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results