accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOBr

CAS RN 10242-11-2

IUPAC Name: 5-bromo-1-benzofuran-2-carboxylic acid
Synonyms: 5-bromo-1-benzofuran-2-carboxylic acid
Molecular Weight (g/mol): 241.04
Molecular Formula: C9H5BrO3
InChi Key: QKUWZCOVKRUXKX-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=CC(Br)=CC=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results