accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 102429-07-2

IUPAC Name: 2-bromo-1-(2,4-difluorophenyl)ethan-1-one
Synonyms: 2-bromo-1-(2,4-difluorophenyl)ethanone
Molecular Weight (g/mol): 235.03
Molecular Formula: C8H5BrF2O
InChi Key: CSGDTHXBRAAOHV-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(C=C1)C(=O)CBr
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)