accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNO

CAS RN 102432-03-1

IUPAC Name: 4-[(1H-imidazol-1-yl)methyl]benzaldehyde
Synonyms: 4-(imidazol-1-ylmethyl)benzaldehyde
Molecular Weight (g/mol): 186.21
Molecular Formula: C11H10N2O
InChi Key: PYVCQHSLEIAQET-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(CN2C=CN=C2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results