accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNO

CAS RN 102432-05-3

IUPAC Name: 3-[(1H-imidazol-1-yl)methyl]benzaldehyde
Synonyms: 3-(imidazol-1-ylmethyl)benzaldehyde
Molecular Weight (g/mol): 186.21
Molecular Formula: C11H10N2O
InChi Key: JMKQKKQFLYPQDD-UHFFFAOYSA-N
SMILES: O=CC1=CC(CN2C=CN=C2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results