accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNSOOClNOO

CAS RN 1024448-59-6

IUPAC Name: 1-benzyl-4-(4-chloro-2-nitrobenzenesulfonyl)piperazine
Synonyms: 1-benzyl-4-(4-chloro-2-nitrobenzenesulfonyl)piperazine
Molecular Weight (g/mol): 395.86
Molecular Formula: C17H18ClN3O4S
InChi Key: ZNLVYSJQUMALEO-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(C=CC(Cl)=C1)S(=O)(=O)N1CCN(CC2=CC=CC=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results