CAS RN 1024598-06-8
CAS RN 1024598-06-8
IUPAC Name:
11-phenyl-11H,12H-indolo[2,3-a]carbazole
Synonyms:
11-phenyl-12H-indolo[2,3-a]carbazole
Molecular Weight (g/mol):
332.41
Molecular Formula:
C24H16N2
InChi Key:
IENOQUQIVSMWGP-UHFFFAOYSA-N
SMILES:
N1C2=CC=CC=C2C2=C1C1=C(C=C2)C2=C(C=CC=C2)N1C1=CC=CC=C1
1
–
1
of
1
results
1
11,12-Dihydro-11-phenylindolo[2,3-a]carbazole 97.0+%, TCI America™

CAS: 1024598-06-8 Molecular Formula: C24H16N2 Molecular Weight (g/mol): 332.41 MDL Number: MFCD22581305 InChI Key: IENOQUQIVSMWGP-UHFFFAOYSA-N Synonym: 11-Phenylindolo[2,3-a]carbazole PubChem CID: 59365871 IUPAC Name: 11-phenyl-11H,12H-indolo[2,3-a]carbazole SMILES: N1C2=CC=CC=C2C2=C1C1=C(C=C2)C2=C(C=CC=C2)N1C1=CC=CC=C1