accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 102507-13-1

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-hydroxy-3-methylbutanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-hydroxy-3-methylbutanoic acid
Molecular Weight (g/mol): 233.26
Molecular Formula: C10H19NO5
InChi Key: SZVRVSZFEDIMFM-ZCFIWIBFSA-N
SMILES: CC(C)(C)OC(=O)N[C@H](C(O)=O)C(C)(C)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results