accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2ONH2O

CAS RN 10255-95-5

IUPAC Name: 2-phenylpropanediamide
Synonyms: 2-phenylpropanediamide
Molecular Weight (g/mol): 178.19
Molecular Formula: C9H10N2O2
InChi Key: CPSUAFUQJBJMPO-UHFFFAOYSA-N
SMILES: NC(=O)C(C(N)=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results