accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1025718-91-5

IUPAC Name: methyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carboxylate
Synonyms: methyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-3-carboxylate
Molecular Weight (g/mol): 263.1
Molecular Formula: C13H18BNO4
InChi Key: FJOJOCXHLGXEQA-UHFFFAOYSA-N
SMILES: COC(=O)C1=CN=CC(=C1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results