accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOONO

CAS RN 102619-05-6

IUPAC Name: 1-(4-methylbenzenesulfonyl)-1H-pyrrole-2-carbaldehyde
Synonyms: 1-(4-methylbenzenesulfonyl)pyrrole-2-carbaldehyde
Molecular Weight (g/mol): 249.28
Molecular Formula: C12H11NO3S
InChi Key: PJJJROQSNDRWIU-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)N1C=CC=C1C=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results