accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONOO

CAS RN 10268-12-9

IUPAC Name: methyl 2-(3-nitrophenyl)acetate
Synonyms: methyl 2-(3-nitrophenyl)acetate
Molecular Weight (g/mol): 195.17
Molecular Formula: C9H9NO4
InChi Key: BFGYITRIATVARH-UHFFFAOYSA-N
SMILES: COC(=O)CC1=CC(=CC=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results