accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3CNSOOFFFFFH2NClRu

CAS RN 1026995-71-0

IUPAC Name: λ²-ruthenium(2+) (2-amino-1,2-diphenylethyl)(2,3,4,5,6-pentafluorobenzenesulfonyl)azanide 1-methyl-4-(propan-2-yl)benzene chloride
Synonyms: λ²-ruthenium(2+) (2-amino-1,2-diphenylethyl)(2,3,4,5,6-pentafluorobenzenesulfonyl)azanide cymene chloride
Molecular Weight (g/mol): 712.14
Molecular Formula: C30H28ClF5N2O2RuS
InChi Key: UWFMZLATRGEOIW-UHFFFAOYNA-M
SMILES: [Cl-].[Ru++].CC(C)C1=CC=C(C)C=C1.NC(C([N-]S(=O)(=O)C1=C(F)C(F)=C(F)C(F)=C1F)C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results