accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10272-07-8

IUPAC Name: 3,5-dimethoxyaniline
Synonyms: 3,5-dimethoxyaniline
Molecular Weight (g/mol): 153.18
Molecular Formula: C8H11NO2
InChi Key: WNRGWPVJGDABME-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=CC(N)=C1
Boiling Point 
  • (2)
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (8)
Quantity 
  • (1)
  • (1)
  • (4)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (2)
  • (5)
  • (3)
  • (2)
  • (8)
  • (1)
  • (1)
  • (4)
  • (3)
  • (1)
  • (2)
  • (3)