CAS RN 10276-09-2
IUPAC Name:
2,2-dimethylbut-3-enoic acid
Synonyms:
3-butenoic acid, 2,2-dimethyl-
Molecular Weight (g/mol):
114.14
Molecular Formula:
C6H10O2
InChi Key:
SCFWAOWWAANBPY-UHFFFAOYSA-N
SMILES:
CC(C)(C=C)C(O)=O
1
–
1
of
1
results
1
Promotions Available
2,2-Dimethyl-3-butenoic acid, 95%
2,2-Dimethyl-3-butenoic acid, 95%
CAS: 10276-09-2 Molecular Formula: C6H10O2 Molecular Weight (g/mol): 114.144 MDL Number: MFCD00196967 InChI Key: SCFWAOWWAANBPY-UHFFFAOYSA-N PubChem CID: 139122 IUPAC Name: 2,2-dimethylbut-3-enoic acid SMILES: CC(C)(C=C)C(=O)O