accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 102783-74-4

IUPAC Name: [({[5-(2-amino-6-oxo-6,9-dihydro-3H-purin-9-yl)-3-hydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy]phosphonic acid
Synonyms: deoxyguanosine diphosphate
Molecular Weight (g/mol): 427.2
Molecular Formula: C10H15N5O10P2
InChi Key: CIKGWCTVFSRMJU-UHFFFAOYNA-N
SMILES: NC1=NC(=O)C2=C(N1)N(C=N2)C1CC(O)C(COP(O)(=O)OP(O)(O)=O)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results