accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 102814-08-4

IUPAC Name: ({[({[(2R,3S,5R)-5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3-hydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy](hydroxy)phosphoryl}oxy)phosphonic acid
Synonyms: dUTP
Molecular Weight (g/mol): 468.14
Molecular Formula: C9H15N2O14P3
InChi Key: AHCYMLUZIRLXAA-SHYZEUOFSA-N
SMILES: O[C@H]1C[C@@H](O[C@@H]1COP(O)(=O)OP(O)(=O)OP(O)(O)=O)N1C=CC(=O)NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results