accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10282-31-2

IUPAC Name: 4-(morpholin-4-yl)benzonitrile
Synonyms: 4-(morpholin-4-yl)benzonitrile
Molecular Weight (g/mol): 188.23
Molecular Formula: C11H12N2O
InChi Key: ZSCUWVQXQDCSRV-UHFFFAOYSA-N
SMILES: N#CC1=CC=C(C=C1)N1CCOCC1
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (3)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)