CAS RN 102830-75-1
IUPAC Name:
3-bromo-5-chloro-2-methoxypyridine
Synonyms:
3-bromo-5-chloro-2-methoxypyridine
Molecular Weight (g/mol):
222.47
Molecular Formula:
C6H5BrClNO
InChi Key:
AYEVELLBDURPJW-UHFFFAOYSA-N
SMILES:
COC1=C(Br)C=C(Cl)C=N1
1
–
1
of
1
results
1
3-Bromo-5-chloro-2-methoxypyridine 98.0+%, TCI America™
CAS: 102830-75-1 Molecular Formula: C6H5BrClNO Molecular Weight (g/mol): 222.466 MDL Number: MFCD08059322 InChI Key: AYEVELLBDURPJW-UHFFFAOYSA-N PubChem CID: 13567751 IUPAC Name: 3-bromo-5-chloro-2-methoxypyridine SMILES: COC1=C(C=C(C=N1)Cl)Br