accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBr

CAS RN 1028648-25-0

IUPAC Name: 9-{[1,1'-biphenyl]-4-yl}-3-(4-bromophenyl)-9H-carbazole
Synonyms: 9-{[1,1'-biphenyl]-4-yl}-3-(4-bromophenyl)carbazole
Molecular Weight (g/mol): 474.4
Molecular Formula: C30H20BrN
InChi Key: AZGVZMFTHCOVID-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results