accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONCH3H3C

CAS RN 10287-53-3

IUPAC Name: ethyl 4-(dimethylamino)benzoate
Synonyms: ethyl-p-dimethylaminobenzoate parbenate ethyl dimethyl paba
Molecular Weight (g/mol): 193.25
Molecular Formula: C11H15NO2
InChi Key: FZUGPQWGEGAKET-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(C=C1)N(C)C
Molecular Weight (g/mol) 
  • (3)
  • (7)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
Quantity 
  • (4)
  • (2)
  • (2)
  • (4)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (6)
  • (3)
  • (7)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (3)
  • (3)
  • (6)
  • (1)