accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CS

CAS RN 102871-31-8

IUPAC Name: 3-pentylthiophene
Synonyms: 3-pentylthiophene
Molecular Weight (g/mol): 154.27
Molecular Formula: C9H14S
InChi Key: PIQKSZYJGUXAQF-UHFFFAOYSA-N
SMILES: CCCCCC1=CSC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results