accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10293-06-8

IUPAC Name: 3-bromo-1,7,7-trimethylbicyclo[2.2.1]heptan-2-one
Synonyms: 3-bromo-d-camphor
Molecular Weight (g/mol): 231.13
Molecular Formula: C10H15BrO
InChi Key: NJQADTYRAYFBJN-UHFFFAOYNA-N
SMILES: CC1(C)C2CCC1(C)C(=O)C2Br
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)