CAS RN 10293-10-4
CAS RN 10293-10-4
IUPAC Name:
3-bromo-7-(bromomethyl)-1,7-dimethylbicyclo[2.2.1]heptan-2-one
Synonyms:
3-bromo-7-(bromomethyl)-1,7-dimethylbicyclo[2.2.1]heptan-2-one
Molecular Weight (g/mol):
310.03
Molecular Formula:
C10H14Br2O
InChi Key:
DCDNKSJBRIJYEC-UHFFFAOYNA-N
SMILES:
CC1(CBr)C2CCC1(C)C(=O)C2Br
1
–
1
of
1
results
1
(+)-3,9-Dibromocamphor 97.0+%, TCI America™
CAS: 10293-10-4 Molecular Formula: C10H14Br2O Molecular Weight (g/mol): 310.029 MDL Number: MFCD00167983 InChI Key: DCDNKSJBRIJYEC-MZIHZIJQSA-N PubChem CID: 13783905 IUPAC Name: (1S,2S,4R)-2-bromo-7-(bromomethyl)-4,7-dimethylbicyclo[2.2.1]heptan-3-one SMILES: CC12CCC(C1(C)CBr)C(C2=O)Br