accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10294-28-7

IUPAC Name: gold(III) bromide
Synonyms: gold(III) bromide
Molecular Weight (g/mol): 439.7
Molecular Formula: AuBr3H3
InChi Key: KJCSJLXVPHOFJC-UHFFFAOYSA-N
SMILES: Br.Br.Br.[Au]
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (3)
  • (2)
Grade 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)